Learn More
Rabbit anti-Sheep IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™
Brand:  Abnova PAB9460.50mg
Product Code. 16214690
  • 997.00 € / 50mg
Specifications
Rabbit anti-Sheep IgM mu heavy chain
Polyclonal
Rabbit anti-sheep IgG recognizes sheep IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide)
Sheep IgM mu heavy chain
50 mg
Sheep
Lyophilized
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Rabbit
This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 5mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.