missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit Lung Thrombomodulin (BDCA-3) Native Protein

Product Code. 11574161
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11574161 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574161 Supplier Invitrogen™ Supplier No. RP43135
  • ONLINE EXCLUSIVE PRICE
    660.00 €
    587.40 € / 50µg
    Save 72.60 €
    11% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Native Protein

RP-43135 contains native Thrombomodulin from rabbit lung. RP-43135 may be used as a positive control in many immunological assays.

Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.2 mg/mL
For Use With (Application) Neutralization
Formulation 0.05% PDOC, TBS with no preservative; pH 7.4
Gene ID (Entrez) 100008887
Molecular Weight (g/mol) 74 kDa
Name Rabbit Lung Thrombomodulin (BDCA-3)
pH Range 7.4
Purification Method SDS-PAGE
Quantity 50 μg
Source Rabbit
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM
Common Name Thrombomodulin (CD141)
Gene Symbol THBD
Product Type Protein
Conjugate Unconjugated
Species Rabbit
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.