missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole sodium, MedChemExpress

Product Code. 16464437
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464437 50 mg 50mg
16448568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16464437 Supplier Medchem Express Supplier No. HYB0656A
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rabeprazole sodium (LY307640 sodium) is a second-generation proton pump inhibitor (PPI) that irreversibly inactivates gastric H+/K+-ATPase. Rabeprazole sodium induces apoptosis. Rabeprazole sodium acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM. Rabeprazole sodium can be used for the research of gastric ulcerations and gastroesophageal reflux.

Rabeprazole sodium (LY307640 sodium) is a second-generation proton pump inhibitor (PPI) that irreversibly inactivates gastric H+/K+-ATPase. Rabeprazole sodium induces apoptosis. Rabeprazole sodium acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM. Rabeprazole sodium can be used for the research of gastric ulcerations and gastroesophageal reflux.

Specifications

Chemical Name or Material Rabeprazole sodium
CAS 117976-90-6
Color White
Purity Grade Notes Research
Molecular Formula C18H20N3NaO3S
Quantity 50 mg
Synonym LY307640 sodium
Solubility Information H2O : ≥ 100 mg/mL (262.18 mM) ∣DMSO : ≥ 48 mg/mL (125.85 mM)
SMILES O=S(C1=NC2=CC=CC=C2N1[Na])CC3=NC=CC(OCCCOC)=C3C
Molecular Weight (g/mol) 381.42
Formula Weight 381.42
Percent Purity 99.09%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.