missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole sodium salt, Thermo Scientific™

Product Code. 17710689 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17710689 5 g 5g
17700689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710689 Supplier Thermo Scientific Acros Supplier No. 470020050
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

Chemical Name or Material Rabeprazole sodium salt
CAS 117976-90-6
Quantity 5 g
Solubility Soluble in chloroform, ethanol, ethylacetate, methanol; inSoluble in ether, hexane
Melting Point 145°C to 155°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C18 H20 N3 Na O3 S
Physical Form Powder
Color White to Yellow to Beige
Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.