missing translation for 'onlineSavingsMsg'
Learn More

rac-1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile, TRC

Product Code. 30404729
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404729 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404729 Supplier Toronto Research Chemicals Supplier No. TRCF595710500MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64169-67-1
Molecular Formula C15 H10 F N O
Quantity 500 mg
Synonym 1-(4-Fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile, 1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile, 1-(4-Fluorophenyl)-5-cyanophthalan, 5-Cyano-1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran, LU 17-046
InChI Formula InChI=1S/C15H10FNO/c16-13-4-2-11(3-5-13)15-14-6-1-10(8-17)7-12(14)9-18-15/h1-7,15H,9H2
SMILES Fc1ccc(cc1)C2OCc3cc(ccc23)C#N
IUPAC Name 1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile
Molecular Weight (g/mol) 239.24
Formula Weight 239.0746
Chemical Name or Material rac-1-(4-Fluoro-phenyl)-1,3-dihydro-isobezofuran-5-carbonitr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.