missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac 1-Oleoyl-2,3-isopropylidieneglycerol-d5, TRC
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | rac 1-Oleoyl-2,3-isopropylidieneglycerol-d5 |
| Molecular Formula | C24 D5 H39 O4 |
| Quantity | 10 mg |
| Synonym | (E)-9-Octadecenoic Acid (2,2-dimethyl-1,3-dioxolan-4-yl)methyl Ester-d5 |
| InChI Formula | InChI=1S/C24H44O4/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(25)26-20-22-21-27-24(2,3)28-22/h11-12,22H,4-10,13-21H2,1-3H3/b12-11+/i20D2,21D2,22D |
| SMILES | [2H]C([2H])(OC(=O)CCCCCCC\C=C\CCCCCCCC)C1([2H])OC(C)(C)OC1([2H])[2H] |
| IUPAC Name | [dideuterio-(4,5,5-trideuterio-2,2-dimethyl-1,3-dioxolan-4-yl)methyl] (E)-octadec-9-enoate |
| Molecular Weight (g/mol) | 401.635 |
| Formula Weight | 401.355 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction