missing translation for 'onlineSavingsMsg'
Learn More

rac-11-Deoxy-8(12)-dehydro Misoprostol, TRC

Product Code. 18063240
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18063240 10 mg 10mg
18023750 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063240 Supplier Toronto Research Chemicals Supplier No. TRCD23263010MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1331639-92-9
Molecular Formula C22 H36 O4
Molecular Weight (g/mol) 364.5188
Synonym (±)-16-Hydroxy-16-methyl-9-oxoprosta-8(12),13-dien-1-oic Acid Methyl Ester, Methyl 7-[2-[4-hydroxy-4-methyloct-1-enyl]-5-oxo-1-cyclopentenyl]heptanoate, (13E)-(+/-)-16-Hydroxy-16-methyl-9-oxoprosta-8(12),13-dien-1-oic Acid Methyl Ester, rac-Misoprostol B1
IUPAC Name methyl 7-[2-(4-hydroxy-4-methyloct-1-enyl)-5-oxocyclopenten-1-yl]heptanoate
SMILES CCCCC(C)(O)CC=CC1=C(CCCCCCC(=O)OC)C(=O)CC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H36O4/c1-4-5-16-22(2,25)17-10-11-18-14-15-20(23)19(18)12-8-6-7-9-13-21(24)26-3/h10-11,25H,4-9,12-17H2,1-3H3
Formula Weight 364.2614
Chemical Name or Material rac-11-Deoxy-8(12)-dehydro Misoprostol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.