missing translation for 'onlineSavingsMsg'
Learn More

rac 2-Aminothiazoline-4-carboxylic Acid, TRC

Product Code. 30011671
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011671 500 mg 500mg
30138393 1 g 1g
30135173 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011671 Supplier Toronto Research Chemicals Supplier No. TRCA630330500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-55-2
Molecular Formula C4 H6 N2 O2 S
Molecular Weight (g/mol) 146.17
Synonym 4-Thiazolecarboxylic acid, 2-amino-4,5-dihydro-, 2-Thiazoline-4-carboxylic acid, 2-amino- (6CI,7CI,8CI), 2-Amino-4,5-dihydro-4-thiazolecarboxylic acid, 2-Amino-2-thiazoline-4-carboxylic acid, 2-Amino-4,5-dihydro-1,3-thiazol-3-ium-4-carboxylate, 2-Amino-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-Aminothiazoline-4-carboxylic acid, 2-Imino-1,3-thiazolidine-4-carboxylic acid, 2-Iminothiazolidine-4-carboxylic acid, DL-2-Amino-2-thiazolin-4-carboxylic acid, DL-2-Amino-δ2-thiazoline-4-carboxylic acid, DL-2-Aminothiazoline-4-carboxylic acid, NSC 25069
IUPAC Name 2-amino-4,5-dihydro-1,3-thiazole-4-carboxylic acid
SMILES NC1=NC(CS1)C(=O)O

Specifications

InChI Formula InChI=1S/C4H6N2O2S/c5-4-6-2(1-9-4)3(7)8/h2H,1H2,(H2,5,6)(H,7,8)
Quantity 500 mg
Formula Weight 146.015
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material rac 2-Aminothiazoline-4-carboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.