missing translation for 'onlineSavingsMsg'
Learn More

rac 2-Chloro Nicotine, TRC

Product Code. 30138062
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138062 25 mg 25mg
30138005 5 mg 5mg
30009658 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138062 Supplier Toronto Research Chemicals Supplier No. TRCC38056525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871894-35-8
Molecular Formula C10H13ClN2
Molecular Weight (g/mol) 196.68
Synonym 2-Chloro-3-(tetrahydro-1-methyl-2-pyrryl)pyridine, 2-Chloro-3-(1-methyl-2-pyrrolidinyl)pyridine
IUPAC Name 2-chloro-3-(1-methylpyrrolidin-2-yl)pyridine
SMILES Fc1cc2[C@@H]3[C@@H](CC=C3)C(Nc2c(F)c1)c4cnccc4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13ClN2/c1-13-7-3-5-9(13)8-4-2-6-12-10(8)11/h2,4,6,9H,3,5,7H2,1H3
Formula Weight 196.0767
Chemical Name or Material rac 2-Chloro Nicotine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.