missing translation for 'onlineSavingsMsg'
Learn More

rac 3-Chloro-1-phenylpropanol, TRC

Product Code. 30135895
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135895 100 mg 100mg
30007105 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135895 Supplier Toronto Research Chemicals Supplier No. TRCC379650100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18776-12-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.64
Synonym alpha-(2-Chloroethyl)benzenemethanol, (-)-3-Chloro-1-phenyl-1-propanol, alpha-(2-Chloroethyl)benzyl Alcohol;
IUPAC Name 3-chloro-1-phenylpropan-1-ol
SMILES OC(CCCl)c1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H11ClO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9,11H,6-7H2
Formula Weight 170.0498
Chemical Name or Material rac 3-Chloro-1-phenylpropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.