missing translation for 'onlineSavingsMsg'
Learn More

rac-3-Hexadecanamido-2-methoxypropan-1-ol Phosphocholine Monohydrate, TRC

Product Code. 30402061
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402061 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402061 Supplier Toronto Research Chemicals Supplier No. TRCH2935005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 163751-35-7
Molecular Formula C25 H51 N2 O6 P . H2 O
Quantity 5 mg
InChI Formula InChI=1S/C25H51N2O6P.H2O/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-25(28)26-22-24(31-5)23-33-34(29,30)32-21-20-27(2,3)4;/h24H,6-13,15-23H2,1-5H3,(H-,26,28,29,30);1H2
SMILES O.CCCCCCCCC[C]CCCCCC(=O)NCC(COP(=O)([O-])OCC[N+](C)(C)C)OC
IUPAC Name [3-(hexadecanoylamino)-2-methoxypropyl] 2-(trimethylazaniumyl)ethyl phosphate;hydrate
Molecular Weight (g/mol) 524.671
Formula Weight 524.359
Chemical Name or Material rac-3-Hexadecanamido-2-methoxypropan-1-ol Phosphocholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.