missing translation for 'onlineSavingsMsg'
Learn More

rac 3-Hydroxy-3-mercaptomethylquinuclidine, TRC

Product Code. 30407130
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407130 25 mg 25mg
30402422 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407130 Supplier Toronto Research Chemicals Supplier No. TRCH94573525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107220-26-8
Molecular Formula C8 H15 N O S
Molecular Weight (g/mol) 173.28
Synonym 3-(Mercaptomethyl)-1-azabicyclo[2.2.2]octan-3-ol, Cevimeline Impurity
IUPAC Name 3-(sulfanylmethyl)-1-azabicyclo[2.2.2]octan-3-ol
SMILES OC1(CS)CN2CCC1CC2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H15NOS/c10-8(6-11)5-9-3-1-7(8)2-4-9/h7,10-11H,1-6H2
Formula Weight 173.0874
Chemical Name or Material rac 3-Hydroxy-3-mercaptomethylquinuclidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.