missing translation for 'onlineSavingsMsg'
Learn More

rac 4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, TRC

Product Code. 30385495
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385495 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385495 Supplier Toronto Research Chemicals Supplier No. TRCD435705100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
CAS 79560-19-3
Molecular Formula C16 H12 Cl2 O
Quantity 100 mg
Synonym 4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one, Sertraline Hydrochloride Imp. F (EP) as Racemate, Sertraline Tetralone Derivative, 1(2H)-Naphthalenone, 4-(3,4-dichlorophenyl)-3,4-dihydro- (9CI), (+)-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, 4-(3,4-Dichlorophenyl)-1-oxo-1,2,3,4-tetrahydronaphthalene, 4-(3,4-Dichlorophenyl)-1-tetralone, 4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, Sertraline tetralone
InChI Formula InChI=1S/C16H12Cl2O/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13/h1-5,7,9,11H,6,8H2
SMILES Clc1ccc(cc1Cl)C2CCC(=O)c3ccccc23
IUPAC Name 4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol) 291.17
Formula Weight 290.0265
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.