missing translation for 'onlineSavingsMsg'
Learn More

rac 4-Hydroxy 3,4-Dihydro-1(2H)-isoquinolinone, TRC

Product Code. 30392324
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392324 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392324 Supplier Toronto Research Chemicals Supplier No. TRCH943560250MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23206-20-4
Molecular Formula C9 H9 N O2
Quantity 250 mg
InChI Formula InChI=1S/C9H9NO2/c11-8-5-10-9(12)7-4-2-1-3-6(7)8/h1-4,8,11H,5H2,(H,10,12)
SMILES OC1CNC(=O)c2ccccc12
IUPAC Name 4-hydroxy-3,4-dihydro-2H-isoquinolin-1-one
Molecular Weight (g/mol) 163.17
Formula Weight 163.0633
Chemical Name or Material rac-3-Hydroxyisoindolin-1-on
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.