missing translation for 'onlineSavingsMsg'
Learn More

rac 7-Methoxy Lasofoxifene, TRC

Product Code. 30477860
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477860 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477860 Supplier Toronto Research Chemicals Supplier No. TRCM26265010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac 7-Methoxy Lasofoxifene
CAS 4796-75-2
Molecular Formula C29H33NO2
Quantity 10 mg
Synonym rel-1-[2-[4-[(1R,2S)-1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl]phenoxy]ethyl]pyrrolidine
InChI Formula InChI=1S/C29H33NO2/c1-31-26-14-16-28-24(21-26)11-15-27(22-7-3-2-4-8-22)29(28)23-9-12-25(13-10-23)32-20-19-30-17-5-6-18-30/h2-4,7-10,12-14,16,21,27,29H,5-6,11,15,17-20H2,1H3/t27-,29+/m1/s1
SMILES COC1=CC2=C(C=C1)[C@H]([C@H](CC2)C3=CC=CC=C3)C4=CC=C(C=C4)OCCN5CCCC5
IUPAC Name 1-[2-[4-[(1R,2S)-6-methoxy-2-phenyl-1,2,3,4-tetrahydronaphthalen-1-yl]phenoxy]ethyl]pyrrolidine
Molecular Weight (g/mol) 427.58
Formula Weight 427.2511
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.