missing translation for 'onlineSavingsMsg'
Learn More

rac (8-Hydroxyquinolin-3-yl)alanine Dihydrochloride, TRC

Product Code. 30392601
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392601 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392601 Supplier Toronto Research Chemicals Supplier No. TRCH85326510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1123191-88-7
Molecular Formula C12 H12 N2 O3 . 2 Cl H
Quantity 10 mg
Synonym 2-Amino-3-(8-hydroxyquinolin-3-yl)propanoic Acid Dihydrochloride, α-Amino-8-hydroxy-3-quinolinepropanoic Acid Hydrochloride, rac HQA
InChI Formula InChI=1S/C12H12N2O3.2ClH/c13-9(12(16)17)5-7-4-8-2-1-3-10(15)11(8)14-6-7;;/h1-4,6,9,15H,5,13H2,(H,16,17);2*1H
SMILES Cl.Cl.NC(Cc1cnc2c(O)cccc2c1)C(=O)O
IUPAC Name 2-amino-3-(8-hydroxyquinolin-3-yl)propanoic acid;dihydrochloride
Molecular Weight (g/mol) 305.16
Formula Weight 304.0381
Chemical Name or Material rac (8-Hydroxyquinolin-3-yl)alanine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.