missing translation for 'onlineSavingsMsg'
Learn More

rac a-Lipoic Acid, TRC

Product Code. 30469072
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469072 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469072 Supplier Toronto Research Chemicals Supplier No. TRCL4687505G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-alpha-Lipoic Acid
CAS 1077-28-7
Purity Grade Notes HPLC
Molecular Formula C8 H14 O2 S2
Quantity 5 g
Synonym Thioctic Acid, 1,2-Dithiolane-3-pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, (±)-1,2-Dithiolane-3-valeric acid, (±)-α-Lipoic acid, (±)-Lipoic acid, (±)-Thioctic acid, (RS)-α-Lipoic acid, (RS)-Lipoic acid, α-Liponic acid, 1,2-Dithiolane-3-valeric acid, 5-(1,2-Dithiolan-3-yl)pentanoic acid, 5-(1,2-Dithiolan-3-yl)valeric acid, 6,8-Thioctic acid, 6-Thioctic acid, Alipure, Biletan, DL-α-Lipoic acid, DL-6,8-Thioctic acid, DL-6-Thioctic acid, DL-Thioctic acid, Espa-lipon, Liposan, Lipothion, NSC 628502, NSC 90788, Protogen A, Pyruvate oxidation factor, Thioctacid, Thioctic acid, Thioctsan, Thiotacid, Tioctacid, Tioctidasi, Tioctidasi acetate replacing factor, dl-Lipoic acid, dl-Thioctic acid
InChI Formula InChI=1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)
SMILES OC(=O)CCCCC1CCSS1
IUPAC Name 5-(dithiolan-3-yl)pentanoic acid
Molecular Weight (g/mol) 206.33
Formula Weight 206.0435
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.