missing translation for 'onlineSavingsMsg'
Learn More

rac Benidipine Hydrochloride, TRC

Product Code. 30135334
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135334 1 g 1g
30134544 100 mg 100mg
30004926 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135334 Supplier Toronto Research Chemicals Supplier No. TRCB1575001G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91599-74-5
Molecular Formula C28 H31 N3 O6 . Cl H
Molecular Weight (g/mol) 542.02
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester, hydrochloride (1:1), (4R)-rel-, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, methyl (3R)-1-(phenylmethyl)-3-piperidinyl ester, monohydrochloride, (4R)-rel- (9CI), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, methyl 1-(phenylmethyl)-3-piperidinyl ester, monohydrochloride, (R*,R*)-(±)-, (±)-Benidipine hydrochloride, Benidipine hydrochloride, Coniel, KW 3049
IUPAC Name 5-O-[(3R)-1-benzylpiperidin-3-yl] 3-O-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES Cl.COC(=O)C1=C(C)NC(=C([C@@H]1c2cccc(c2)[N+](=O)[O-])C(=O)O[C@@H]3CCCN(Cc4ccccc4)C3)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H31N3O6.ClH/c1-18-24(27(32)36-3)26(21-11-7-12-22(15-21)31(34)35)25(19(2)29-18)28(33)37-23-13-8-14-30(17-23)16-20-9-5-4-6-10-20;/h4-7,9-12,15,23,26,29H,8,13-14,16-17H2,1-3H3;1H/t23-,26-;/m1./s1
Formula Weight 541.198
Chemical Name or Material Benidipine, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.