missing translation for 'onlineSavingsMsg'
Learn More

rac Carprofen, TRC

Product Code. 30137758
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137758 1 g 1g
30040646 50 mg 50mg
30040896 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137758 Supplier Toronto Research Chemicals Supplier No. TRCC1843501G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53716-49-7
Molecular Formula C15 H12 Cl N O2
Molecular Weight (g/mol) 273.71
Synonym 9H-Carbazole-2-acetic acid, 6-chloro-α-methyl-, (±)-, 6-Chloro-α-methyl-9H-carbazole-2-acetic acid, (dl)-6-Chloro-α-methylcarbazole-2-acetic acid, 2-(6-Chlorocarbazol-2-yl)propionic acid, 6-Chloro-α-methylcarbazole-2-acetic acid, C 5720, Carprodyl, Carprofen, Imadyl, NSC 297935, Rimadyl, Ro 20-5720, Ro 20-5720/000
IUPAC Name 2-(6-chloro-9H-carbazol-2-yl)propanoic acid
SMILES CC(C(=O)O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)
Formula Weight 273.0557
Chemical Name or Material Carprofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.