missing translation for 'onlineSavingsMsg'
Learn More

rac cis-3-((tert-Butyldimethylsilyl)oxy)-1-(4-nitrophenyl)cyclopentanecarboxamide, TRC

Product Code. 30330545
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330545 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330545 Supplier Toronto Research Chemicals Supplier No. TRCB69226510MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac cis-3-((tert-Butyldimethylsilyl)oxy)-1-(4-nitrophenyl)cyclopentanecarboxamide
CAS 2518265-03-5
Molecular Formula C18 H28 N2 O4 Si
Quantity 10 mg
Synonym (1S,3S)-3-((tert-Butyldimethylsilyl)oxy)-1-(4-nitrophenyl)cyclopentanecarboxamide
InChI Formula InChI=1S/C18H28N2O4Si/c1-17(2,3)25(4,5)24-15-10-11-18(12-15,16(19)21)13-6-8-14(9-7-13)20(22)23/h6-9,15H,10-12H2,1-5H3,(H2,19,21)/t15-,18-/m0/s1
SMILES CC(C)(C)[Si](C)(C)O[C@H]1CC[C@@](C1)(C(=O)N)c2ccc(cc2)[N+](=O)[O-]
IUPAC Name (1S,3S)-3-[tert-butyl(dimethyl)silyl]oxy-1-(4-nitrophenyl)cyclopentane-1-carboxamide
Molecular Weight (g/mol) 364.511
Formula Weight 364.182
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.