missing translation for 'onlineSavingsMsg'
Learn More

rac-Clopidogrel Carboxylic Acid Hydrochloride, TRC

Product Code. 30367904
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367904 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367904 Supplier Toronto Research Chemicals Supplier No. TRCC58722025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Clopidogrel Carboxylic Acid Hydrochloride
CAS 1015247-88-7
Molecular Formula C15H15Cl2NO2S
Quantity 25 mg
Synonym rac-2-(2-Chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic Acid Hydrochloride, rac-α-(2-Chlorophenyl)-6,7-dihydro-thieno[3,2-c]pyridine-5(4H)-acetic Acid Hydrochloride
InChI Formula InChI=1S/C15H14ClNO2S.ClH/c16-12-4-2-1-3-11(12)14(15(18)19)17-7-5-13-10(9-17)6-8-20-13;/h1-4,6,8,14H,5,7,9H2,(H,18,19);1H
SMILES ClC(C=CC=C1)=C1C(C(O)=O)N2CCC3=C(C=CS3)C2.Cl
IUPAC Name 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid;hydrochloride
Molecular Weight (g/mol) 344.26
Formula Weight 343.0201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.