missing translation for 'onlineSavingsMsg'
Learn More

rac Dihydro Mebendazole, TRC

Product Code. 30135607
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135607 500 mg 500mg
30004188 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135607 Supplier Toronto Research Chemicals Supplier No. TRCD449320500MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60254-95-7
Molecular Formula C16 H15 N3 O3
Molecular Weight (g/mol) 297.31
Synonym Carbamic acid, [5-(hydroxyphenylmethyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), R 19167
IUPAC Name methyl N-[5-[hydroxy(phenyl)methyl]-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)C(O)c3ccccc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H15N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9,14,20H,1H3,(H2,17,18,19,21)
Formula Weight 297.1113
Percent Purity >95%
Chemical Name or Material Mebendazole-5-hydroxy
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.