missing translation for 'onlineSavingsMsg'
Learn More

rac Dobutamine-d4 Hydrochloride, TRC

Product Code. 30381356
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381356 1 mg
30371642 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381356 Supplier Toronto Research Chemicals Supplier No. TRCD4944471MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246815-74-6
Molecular Formula C18 2H4 H19 N O3 . Cl H
Molecular Weight (g/mol) 341.87
Synonym 4-[2-[[3-(4-Hydroxyphenyl)-1-methylpropyl]amino]ethyl-1,1,2,2-d4]-1,2-benzenediol hydrochloride, Dobutamine-d4 hydrochloride, Dobutamine-D4 hydrochloride (aminoethyl-1,1,2,2-D4)
IUPAC Name 4-[1,1,2,2-tetradeuterio-2-[4-(4-hydroxyphenyl)butan-2-ylamino]ethyl]benzene-1,2-diol;hydrochloride
SMILES Cl.[2H]C([2H])(NC(C)CCc1ccc(O)cc1)C([2H])([2H])c2ccc(O)c(O)c2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-7-16(20)8-5-14)19-11-10-15-6-9-17(21)18(22)12-15;/h4-9,12-13,19-22H,2-3,10-11H2,1H3;1H/i10D2,11D2;
Formula Weight 341.1696
Chemical Name or Material Dobutamine aminoethyl-d4 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.