missing translation for 'onlineSavingsMsg'
Learn More

rac Galaxolidone (Mixture of Diastereomers), TRC

Product Code. 17991515
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991515 50 mg 50mg
17900495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991515 Supplier Toronto Research Chemicals Supplier No. TRCG18900550MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507442-49-1
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.38
Synonym 4,6,7,8-Tetrahydro-4,6,6,7,8,8-hexamethyl-cyclopenta[g]-2-benzopyran-1(3H)-one
IUPAC Name 4,6,6,7,8,8-hexamethyl-4,7-dihydro-3H-cyclopenta[g]isochromen-1-one
SMILES O=C1OCC(C)C2=CC3=C(C(C)(C)C(C)C3(C)C)C=C21

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H24O2/c1-10-9-20-16(19)13-8-15-14(7-12(10)13)17(3,4)11(2)18(15,5)6/h7-8,10-11H,9H2,1-6H3
Formula Weight 272.1776
Percent Purity >95%
Chemical Name or Material rac Galaxolidone (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.