missing translation for 'onlineSavingsMsg'
Learn More

rac Ketoprofen, TRC

Product Code. 30392467
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392467 Supplier Toronto Research Chemicals Supplier No. TRCK2008005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ketoprofen
CAS 22071-15-4
Purity Grade Notes HPLC
Molecular Formula C16 H14 O3
Quantity 5 g
Synonym Benzeneacetic acid, 3-benzoyl-alpha-methyl- (9CI), Hydratropic acid, m-benzoyl- (8CI), (+/-)-2-(3-Benzoylphenyl)propionic acid, (+/-)-3-Benzoyl-alpha-methylbenzeneacetic acid, (+/-)-Ketoprofen, (+/-)-m-Benzoylhydratropic acid, (RS)-Ketoprofen, alpha-(3-Benzoylphenyl)propionic acid, 19583RP, 2-(3-Benzoylphenyl)propionic acid, 2-(m-Benzoylphenyl)propionic acid, 3-Benzoyl-alpha-methylbenzeneacetic acid, 3-Benzoylhydratropic acid, Alrheumat, Alrheumun, Aneol, Bi-profenid, Capisten, Dexal, Epatec, Fastum, Iso-K, Kefenid, Ketoprofen, Ketoprofene, Ketopron, Ketoprophen, Ketorin, Knavon, Lertus, Menamin, Meprofen, Miltax, Mohrus, Orudis, Orugesic, Oruvail, Oscorel, Profenid, R.P. 19583, RU 4733, Racemic ketoprofen, Toprec, Toprek, m-Benzoylhydratropic acid
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)
SMILES CC(C(=O)O)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name 2-(3-benzoylphenyl)propanoic acid
Molecular Weight (g/mol) 254.28
Formula Weight 254.0943
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.