missing translation for 'onlineSavingsMsg'
Learn More

rac Lenalidomide-13C5, TRC

Product Code. 30481180
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481180 10 mg 10mg
30479463 1 mg 1mg
30477065 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481180 Supplier Toronto Research Chemicals Supplier No. TRCL32800210MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219332-91-8
Molecular Formula C813C5H13N3O3
Molecular Weight (g/mol) 264.22
Synonym Lenalidomide 13C5
SMILES Nc1cccc2C(=O)N(Cc12)[13CH]3[13CH2][13CH2][13C](=O)N[13C]3=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H13N3O3/c14-9-3-1-2-7-8(9)6-16(13(7)19)10-4-5-11(17)15-12(10)18/h1-3,10H,4-6,14H2,(H,15,17,18)/i4+1,5+1,10+1,11+1,12+1
Chemical Name or Material rac Lenalidomide-13C5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.