missing translation for 'onlineSavingsMsg'
Learn More

rac N-Demethyl Promethazine Hydrochloride, TRC

Product Code. 30008442
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008442 250 mg 250mg
17941585 25 mg 25mg
17963015 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008442 Supplier Toronto Research Chemicals Supplier No. TRCD231220250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60113-77-1
Molecular Formula C16 H18 N2 S . Cl H
Molecular Weight (g/mol) 306.85
Synonym (2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride, Promethazine Hydrochloride Imp. C (EP) as Hydrochloride, Norpromethazine Hydrochloride, 10H-Phenothiazine-10-ethanamine, N,α-dimethyl-, hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,α-dimethyl-, monohydrochloride (9CI)
IUPAC Name N-methyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
SMILES Cl.CNC(C)CN1c2ccccc2Sc3ccccc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H18N2S.ClH/c1-12(17-2)11-18-13-7-3-5-9-15(13)19-16-10-6-4-8-14(16)18;/h3-10,12,17H,11H2,1-2H3;1H
Formula Weight 306.0957
Percent Purity >95%
Chemical Name or Material (2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride (Norpromethazine Hydrochloride)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.