missing translation for 'onlineSavingsMsg'
Learn More

rac-Nicotine-1’-oxide, TRC

Product Code. 30478126
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478126 100 mg 100mg
30464884 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478126 Supplier Toronto Research Chemicals Supplier No. TRCN427490100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Nicotine-1'-oxide
Purity Grade Notes HPLC
Molecular Formula C10H14N2O
Quantity 100 mg
Synonym Racemic Mixture of (S,S)-Nicotine-1’-oxide and (R,R)-Nicotine-1’-oxide
SMILES C[N@@+](CCC1)([O-])[C@@H]1C2=CC=CN=C2
Molecular Weight (g/mol) 178.23
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.