missing translation for 'onlineSavingsMsg'
Learn More

rac Pimobendan, TRC

Product Code. 30408967
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408967 5 mg -
30410677 50 mg -
30428746 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408967 Supplier Toronto Research Chemicals Supplier No. TRCP4475005MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74150-27-9
Molecular Formula C19 H18 N4 O2
Molecular Weight (g/mol) 334.37
Synonym (5RS)-6-[2-(4-Methoxyphenyl)-1H-benzimidazol-5-yl]-5-methyl-4,5-dihydropyridazin-3(2H)-one, 3(2H)-Pyridazinone, 4,5-dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-, 3(2H)-Pyridazinone, 4,5-dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-5-methyl- (9CI), 4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone, Acardi, Pimobendan, Racemic pimobendan, UD-CG 115, UD-CG 115BS, Vetmedin, dl-Pimobendan
IUPAC Name 3-[2-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-4-methyl-4,5-dihydro-1H-pyridazin-6-one
SMILES COc1ccc(cc1)c2nc3cc(ccc3[nH]2)C4=NNC(=O)CC4C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H18N4O2/c1-11-9-17(24)22-23-18(11)13-5-8-15-16(10-13)21-19(20-15)12-3-6-14(25-2)7-4-12/h3-8,10-11H,9H2,1-2H3,(H,20,21)(H,22,24)
Formula Weight 334.143
Chemical Name or Material Pimobendan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.