missing translation for 'onlineSavingsMsg'
Learn More

(rac,syn)-4-Deschloro-sertraline Hydrochloride, TRC

Product Code. 30367156
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367156 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367156 Supplier Toronto Research Chemicals Supplier No. TRCD2896751MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,S)-4-Deschloro-sertraline hydrochloride
CAS 871838-58-3
Molecular Formula C17 H18 Cl N . Cl H
Quantity 1 mg
Synonym Sertraline EP Impurity D HCl
InChI Formula InChI=1S/C17H18ClN.ClH/c1-19-17-10-9-14(12-5-4-6-13(18)11-12)15-7-2-3-8-16(15)17;/h2-8,11,14,17,19H,9-10H2,1H3;1H/t14-,17-;/m0./s1
SMILES Cl.CN[C@H]1CC[C@@H](c2cccc(Cl)c2)c3ccccc13
IUPAC Name (1S,4S)-4-(3-chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
Molecular Weight (g/mol) 308.25
Formula Weight 307.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.