missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Radiation Safety Beta Containers

Product Code. 12387137 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W x H):
490 x 550 x 370mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12387137 490 x 550 x 370mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12387137
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Featuring sizeable ergonomic working space and a sitting or standing position for unobstructed views; these unqiue Cleaver Scientific cabinets provide a convenient area to carry out work with beta and gamma emitting isotopes with complete all round protection.

  • Cleaver Scientific cabinets provide a convenient area to carry out work with beta and gamma emitting isotopes with complete all round protection
  • Each work cabinet's 49 x 55 x 37cm dimensions offer a large working area without impeding vision, either in a standing or seated position
  • Services can also be used inside the cabinet via a rear port
  • The back panel has a pipette holder to keep the work surface clear
  • The unit can store radioactive materials when the hinged doors are closed
  • These cabinets are compatible with 68 x 54cm trays, CSR-TY4, CSR-TW4 and CSR-TO4
  • Sizeable ergonomic working space, option of being fully enclosed, sitting or standing position for unobstructed views, all round user protection

Specifications

Dimensions (L x W x H) 490 x 550 x 370mm
For Use With (Application) Beta and gamma emitting isotopes
Product Type Radiation Safety Containers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.