missing translation for 'onlineSavingsMsg'
Learn More

Ramipril, TRC

Product Code. 30413811
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413811 1 g 1g
30419905 100 mg 100mg
30411708 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30413811 Supplier Toronto Research Chemicals Supplier No. TRCR1110001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Purity >95% (HPLC)
CAS 87333-19-5
Molecular Weight (g/mol) 416.51
Quantity 1 g
Shipping Condition Room Temperature
Synonym Ramipril∣(2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid∣(2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid∣ [2S-[1[R*(R*)],2α,3aβ,6aβ]]-1-[2-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid∣Altace∣Cardace∣Delix∣HOE 498∣Hopace∣Pramace∣Quark∣Ramace∣Ramipres∣Ramipril∣Triatec∣Tritace∣Unipril∣Vesdil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.