missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Streptavidin-HRP

Product Code. 18794494
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18794494 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18794494 Supplier R&D Systems™ Supplier No. DY998
  • ONLINE EXCLUSIVE PRICE
    374.45 €
    332.85 € / Pack of 5
    Save 41.60 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in ELISAs, dilute to the working concentration specified on the vial label using 1% BSA/PBS or Reagent Diluent Concentrate 2 (Catalog # DY995) diluted 1:10 in deionized water.

Stable for up to 6 months after opening when stored at 2° to 8°C
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 1 mL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
For Use With (Application) ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.