missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Random Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems
Shop All Applied Biosystems ProductsDescription
- This Random Access Storage Option is designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput.
- This storage option holds up to 24 plates/3 hotels (384-well). The hotels enable users to easily add new plates or change the order in which the plates are run. Hotels are ideal when there are multiple users using 384-well plates. This option is not ideal for 96-well plates.
- Couple this Random Access Storage Option with the Orbitor RS2 Microplate Mover for QuantStudio Systems to complete your automation bundle.
- For integration of two QuantStudio systems with the Orbitor RS2 mover, the Dual QuantStudio System Integration Kit is required in addition to the automation bundle.
Specifications
Specifications
| For Use With (Equipment) | QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Room Temperature |
| Description | Storage Option |
| Quantity | 1 each |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?