missing translation for 'onlineSavingsMsg'
Learn More

Raney™-Nickel, Activated Catalyst, 50% slurry in water

Product Code. 10243353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10243353 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10243353 Supplier Thermo Scientific Acros Supplier No. 395921000
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0 , 7732-18-5 , 7429-90-5
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 100 g
Packaging Plastic bottle
Form Slurry
Formula Weight 58.69
Chemical Name or Material Raney™-Nickel, Activated Catalyst
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Raney Nickel, activated catalyst, 50% slurry in water
Signal Word
  • Danger
Hazard Category
  • Self-heating substance Category 1
  • Specific target organ toxicity Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H251-Self-heating: may catch fire.
  • H372-Causes damage to organs.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P235+P410-Keep cool. Protect from sunlight.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
  • MIXTURE LIST-Contain: Nickel, Aluminium powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.