missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Raphin 1
GREENER_CHOICE

Product Code. 18779936
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18779936

Brand: Tocris Bioscience™ 6760/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of the regulatory subunit PPP1R15B of protein phosphatase 1 (Kd = 33 nM); orally bioavailable and brain penetrant

Raphin 1 is an inhibitor of the regulatory subunit PPP1R15B of protein phosphatase (Kd = 33 nM). Displays >30-fold selectivity for PPP1R15B over PPP1R15A. Inhibits PPP1R15B-mediated dephosphorylation and protein synthesis in vitro. Induces transient reduction in protein synthesis in brains of wild type mice after single dose. Decreases SDS-insoluble huntingtin assemblies and nuclear inclusions in a mouse model of Huntington's disease. Orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-2-[(2,3-Dichlorophenyl)methylene]hydrazinecarboximidamide
CAS 2022961-17-5
Quantity 10 mg
Molecular Formula C8H8Cl2N4
Formula Weight 231.08
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.