missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Rapid H. pylori Antigen Test Card

Product Code. 30280612 Shop All MP Biomedicals Products
Change view
Click to view available options
Clia Complexity:
Not Applicable
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Clia Complexity unitSize
30280612 Not Applicable 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280612 Supplier MP Biomedicals Supplier No. 07HPY025ST

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapid H. pylori Antigen Test Card is used for the qualitative detection of Helicobacter pylori antigens in human fecal samples.

  • Rapid H. pylori Antigen Test Card is an immunochromatographic membrane assay that uses highly sensitive antibodies to detect H. pylori antigens in stool samples.
  • The reagent pad of the test strip contains red-colored colloidal gold conjugated with antibodies against H. pylori antigens.
  • The reaction membrane contains the secondary antibodies for H.pylori antigens, which are pre-immobilized in the test line.
  • This test is only to be used by healthcare professionals. It cannot be used alone as the basis for the diagnosis and exclusion of cases.
  • CE-marked
  • Accurate result within 10 minutes
  • One-step procedure
  • No cross-reaction
  • High sensitivity
  • For professional use

Especificaciones

CE Marker Self-declared
Clia Complexity Not Applicable
Detectable Analytes H. pylori Antigen
Sample Type Human stool sample
Sensitivity 99.00%
Type Rapid Test
Disposable Single-use
Form Ready-to-Use (RTU)
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.