missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
- The Rapid Influenza A+B Antigen Test Card is an immunochromatographic membrane assay that uses highly sensitive monoclonal antibodies to detect influenza type A and B nucleoprotein antigens in nasopharyngeal swabs and nasal aspirate samples.
- The reagent pad of the test strip contains the colloidal-gold conjugated with the monoclonal antibodies against Influenza virus A and B; the reaction membrane contains the secondary antibodies for virus A or for B, and the polyclonal antibodies against the mouse globulin, which are pre-immobilized on the membrane.
- This test is only to be used by healthcare professionals, not for home testing. It cannot be used alone as the basis for the diagnosis and exclusion of cases.
- Accurate Result within 10 minutes
- One-step procedure, No equipment needed
- No cross-reaction
- High sensitivity, specificity, and accuracy
- CE marked
- For professional use only
- This test is only to be used by healthcare professionals, not for home testing. It cannot be used alone as the basis for the diagnosis and exclusion of cases.
Specifica
Specifica
| CE Marker | Self-declared |
| Clia Complexity | Not Applicable |
| Detectable Analytes | Influenza A, Influenza B |
| Detection Method | Immunochromatographic lateral flow |
| Format | Test card |
| Sample Type | Human nasopharyngeal swabs, Human nasal swabs, Human nasal aspirate samples |
| Sensitivity | Influenza A: 1.2 x 103 TCID{50}/mL, Influenza B: 1.0 x 104 TCID{50}/mL |
| Type | Rapid Test |
| For Use With (Application) | Qualitative detection of influenza type A and type B nucleoprotein antigens in nasopharyngeal swabs, nasal swabs, and nasal aspirate samples |
| Form | Ready-to-Use (RTU) |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?