missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Rapid Influenza A Test Card (Flu A)

Product Code. 30280599 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Influenza
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280599 Influenza 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280599 Supplier MP Biomedicals Supplier No. 07FLUA020
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid Influenza A Test Card (Flu A) is used for the qualitative detection of influenza type A nucleoprotein antigens in nasopharyngeal swabs, nasal swabs, and nasal aspirate samples.

  • Rapid Influenza A Antigen Test Card utilizes the principle of immuno-chromatography.
  • Mouse anti-influenza A are immobilized on the nitrocellulose membrane, as one individual test line (T line) in the test window of the test device.
  • As the test sample flows through the membrane within the test device, the colored mouse anti-influenza A antibodies-colloidal gold conjugate forms complexes with influenza A viral antigens, if present in the sample.
  • This complex moves further on the membrane to the test region where it is captured by the mouse anti-influenza A antibodies coated on the membrane leading to formation of a colored band, which indicates positive test results.
  • Absence of this colored band in the test window indicates a negative test result.
  • A built-in control line will always appear in the test window when the test is performed properly, regardless of the presence or absence of influenza A viral antigens in the specimen.
  • Accurate Result within 15 minutes
  • One-step procedure, No equipment needed
  • No cross-reaction
  • High sensitivity, specificity, and accuracy
  • CE marked
  • For professional use only
  • This test is only to be used by healthcare professionals, not for home testing. It cannot be used alone as the basis for the diagnosis and exclusion of cases.

Specifications

CE Marker Self-declared
Clia Complexity Not Applicable
Detectable Analytes Influenza
Detection Method Immunochromatographic lateral flow
Format Test card
Sample Type Human nasopharyngeal swab, Human nasal swab, Human nasal aspirate samples
Sensitivity 1.2 x 103 TCID{50}/mL
Type Rapid Test
For Use With (Application) Qualitative detection of Influenza type A nucleoprotein antigens in human nasopharyngeal swabs, nasal swabs, and nasal aspirate samples
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.