missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Rapid Salmonella typhi IgG/IgM Combo Test Card

Product Code. 30280610 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Salmonella Antigen
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280610 Salmonella Antigen 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280610 Supplier MP Biomedicals Supplier No. 07SAL025AB
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid Salmonella typhi IgG/IgM Combo Test Card is used for the qualitative detection of Salmonella typhi antibodies in human whole blood, serum or plasma.

  • Rapid Salmonella typhi IgG/IgM Combo Test Card is an immunochromatographic membrane assay.
  • It is designed for the rapid qualitative determination of IgG and IgM antibodies to Salmonella typhi in human serum, plasma, or whole blood.
  • The S.typhi antigens-colloidal gold conjugated combine with specific antibodies (IgG and/or IgM) of S.typhi, if present in the sample.
  • These complexes move to the test regions by capillarity and are captured by anti-S. typhi IgG and/or IgM antibodies pre-immobilized in the membrane, leading to the formation of colored band and indicating a positive test result.
  • This test is only to be used by healthcare professionals. It cannot be used alone as the basis for the diagnosis and exclusion of cases.
  • CE-marked
  • Accurate Result within 20 minutes
  • One-step procedure
  • For professional use

Specifications

CE Marker Self-declared
Clia Complexity Not Applicable
Detectable Analytes Salmonella Antigen
Format Test card
Sample Type Human whole blood, Human serum, Human plasma
Sensitivity Determined by chemiluminescence method: 2.38 (S/CO value) for Salmonella typhi IgG 2.57 (S/CO value) for Salmonella typhi IgM
Type Rapid Test
For Use With (Application) For the qualitative detection of Salmonella typhi antibodies in human whole blood, serum or plasma. The test is intended to aid in the rapid diagnosis of S. typhi infection, responsible for typhoid fever.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.