missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Rapid SARS-CoV-2 Antigen Test

The Rapid SARS-CoV-2 Antigen Test Card is an in-vitro diagnostic test for private use. It is used to detect SARS-CoV-2 antigens from anterior nasal swabs.

Brand:  MP Biomedicals 07AG6005BS-DE-EN-FR

Product Code. 17237746

  • ONLINE EXCLUSIVE PRICE
    23.05 €
    15.39 € / 5 tests
    Save 7.66 €
    33% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
This item is not returnable. View return policy

Description

Description

  • Easy sampling using anterior nasal swab specimens
  • Results in 15 minutes
  • Works with a nasal swab of 2.5 cm
  • Specificity 99.20%, Sensitivity 96.77%, Accuracy 98.72
  • Includes all required consumable materials
  • No cross-contamination with other coronaviruses like MERS, 229E, NL63, OC43 or influenza A, or B

Usage Statement: Test for self-testing by laypersons with CE certification

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.