missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Rapid V. Cholerae Test O139

Product Code. 30280613 Shop All MP Biomedicals Products
Change view
Click to view available options
Reaction Speed:
10 min.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Reaction Speed unitSize
30280613 10 min. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280613 Supplier MP Biomedicals Supplier No. 07VCH139C
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapid V. Cholerae Tests O139 are used for the qualitative detection of Vibrio cholerae serogroup O139 in human fecal specimens, environmental water or solid food specimens.

  • Rapid V. Cholerae O139 Test is an immunochromatographic membrane assay that uses highly sensitive antibodies to detect V. cholerae antigens in stool sample, environmental water or solid food specimens.
  • The reagent pad of the test strip contains colored colloidal gold conjugated with monoclonal antibodies against V. cholerae O139 antigens.
  • The reaction membrane contains the secondary antibodies for V. cholerae O139 antigens, which are pre-immobilized in the test line.
  • This test is only to be used by healthcare professionals. It cannot be used alone as the basis for the diagnosis and exclusion of cases.
  • CE-marked
  • Accurate result within 10 minutes
  • One-step procedure
  • No cross-reaction
  • For professional use

Specifications

Disease Cholera
Form Test card
Detection Method Rapid Detection
Sample Type Environmental Sample, Food Sample
Species Vibrio cholerae serougroup O139
Reaction Speed 10 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.