missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat 4-1BB Ligand/TNFSF9 Protein

Product Code. 18716736
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18716736

Brand: R&D Systems™ 91884L025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Rat 4-1BB/TNFRSF9 Fc Chimera (Catalog # 7968-4B ) is Immobilized at 100 ng/mL (100 μL/well), the concentration of Recombinant Rat 4-1BB Ligand/TNFSF9 that produces 50% optimal binding response is approximately 1-6 ng/mL. Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for for this effect is 5-30 ng/mL

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q80WE6
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 8744
Molecular Weight (g/mol) M.W. (Observed): 33-43 kDa, reducing conditions; M.W. (theoretical): 24 kDa
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived rat 4-1 BB Ligand/TNFSF9 protein Arg103-Gly308, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Biological Activity 5 to 30ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein 4-1 BB Ligand/TNFSF9
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.