missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Rat Alb native Protein from rat serum

Product Code. 16222280
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16222280 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16222280 Supplier Abnova™ Supplier No. P5371.100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

Specifications

For Use With (Application) SDS-PAGE
Gene ID (Entrez) 24186
Name Alb (Rat) Native Protein
Preparation Method Native protein from rat serum, fraction V
Quantity 100 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Gene Alias Alb1/Albza
Common Name Alb
Gene Symbol Alb
Species Rat
Protein Tag None
Expression System Native protein from rat serum, fraction V
Form Lyophilized
Purity or Quality Grade 100% by agarose electrophoresis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.