missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Rat anti-Mouse, Clone: 37801/37875, R&D Systems™

Rat Monoclonal Antibody has been used in 18 publications
Brand: R&D Systems MAB785-100
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| IFN-gamma | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFNG | |
| E. coli-derived recombinant mouse IFN-gamma | |
| 100 μg | |
| Primary | |
| Detects mouse IFN-gamma in ELISAs. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) C10, rmG-CSF, rmGM-CSF, recombinant human IFN-gamma , rmIL-1 alpha , rmIL-1 beta , rmIL-2, rmIL-3, rmIL-4, rmIL-5, rmIL-6, rmIL-7, rmIL-9, rmIL-10, rmIL-10 R, rmIL-12, rmIL-13, rmJE, rmKC, rmLIF, rmM-CSF, rmMIP-1 alpha , rmMIP-1 beta , rmMIP-2, rmSCF, rmTNF-alpha , rmTpo, and rmVEGF. | |
| Mouse | |
| Supernatant |
| ELISA | |
| 37801/37875 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3458 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a, IgG2B Cocktail |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?