missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgE, Clone: R35-72, Secondary Antibody, BD

Product Code. 10628810
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10628810 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10628810 Supplier BD Biosciences Supplier No. 553413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The rat anti-mouse IgE antibody (clone R35-72) reacts specifically with mouse IgE of the Igh-C [a] and Igh-C [b] haplotypes. It has been reported not to react with other Ig isotypes. Detection with the rat anti-mouse IgE antibody (clone R35-72) of surface

Host Species: Rat
Clone: R35-72
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Mouse IgE (pooled)

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgE
Classification Monoclonal
Clone R35-72
Concentration 0.5mg/mL
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.