missing translation for 'onlineSavingsMsg'
Learn More

Rat Anti-Mouse IgG1, Clone: X56, Secondary Antibody, BD

Product Code. 15816418
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15816418 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15816418 Supplier BD Biosciences Supplier No. 550874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The X56 antibody reacts specifically with mouse IgG1 of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes.

Host Species: Rat
Clone: X56
Isotype: IgG1 κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Anti-Mouse IgG1
Classification Monoclonal
Clone X56
Concentration 0.2mg/mL
Conjugate APC
Host Species Rat
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.