Learn More
Rat anti-Mouse IgG2b Heavy Chain Secondary Antibody (LO-MG2b-2), Janelia Fluor 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-68438JF646
Product Code. 30232527
  • 702.00 € / 0.25mL
Description
IgG2b Heavy Chain Monoclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG2b Heavy Chain
Monoclonal
Janelia Fluor 646
Rat
Protein A purified
RUO
Mouse
Purified
ELISA
LO-MG2b-2
50mM Sodium Borate
Purified mouse IgG2b
0.25 mL
Secondary
Store at 4°C in the dark.
IgG1