missing translation for 'onlineSavingsMsg'
Learn More

Rat Anti-Mouse IgM, Clone: II/41, Secondary Antibody, BD

Product Code. 15806368
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15806368 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15806368 Supplier BD Biosciences Supplier No. 550676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation.

Host Species: Rat
Clone: II/41
Isotype: IgG2a κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Anti-Mouse IgM
Classification Monoclonal
Clone II/41
Concentration 0.2mg/mL
Conjugate APC
Host Species Rat
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.