missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgM, Clone: R6-60.2, Secondary Antibody, BD
Description
The R6-60.2 antibody reacts specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. R6-60.2 antibody has not been shown to stimulate B-cell proliferation. This antibody is routinely tested by flow cytomet
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Rat anti-Mouse IgM |
| Classification | Monoclonal |
| Clone | R6-60.2 |
| Concentration | 0.5mg/mL |
| Host Species | Rat |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?